Learn More
Cayman Chemical 4-Methylumbelliferyl-2-acetamido-2-deoxy-β-D-Glucopyranoside, Purity: ≥98%, CAS: 37067-30-4, 100 mg
SDP
Catalog No. 502941332
Encompass_Preferred
A fluorogenic substrate for β-hexosaminidases; is cleaved by β-hexosaminidases to release the fluorescent moiety 4-ΜU; ex = 320 and 360 nm at low (1.97-6.72) and high pH (7.12-10.3), respectively; em = 445 to 455 nm, increasing as pH decreases; has been used to quantify β-hexosaminidase activity in serum or leukocytes from patients with GM2 gangliosidoses such as Tay-Sachs disease
Catalog No. 50-294-1332 Supplier Cayman Chemical Supplier No. 26953100 MG
A fluorogenic substrate for β-hexosaminidases; is cleaved by β-hexosaminidases to release the fluorescent moiety 4-ΜU; ex = 320 and 360 nm at low (1.97-6.72) and high pH (7.12-10.3), respectively; em = 445 to 455 nm, increasing as pH decreases; has been used to quantify β-hexosaminidase activity in serum or leukocytes from patients with GM2 gangliosidoses such as Tay-Sachs disease