Learn More
Cayman Chemical 4-Methylumbelliferyl-α-D-Galactopyranoside, Purity: ≥98%, CAS: 38597-12-5, 100 mg
SDP
Catalog No. 502968203
Encompass_Preferred
A fluorogenic substrate of α-galactosidase
Catalog No. 50-296-8203 Supplier Cayman Chemical Supplier No. 16551100 MG
A fluorogenic substrate of α-galactosidase