Learn More
Cayman Chemical 4MethyLMbelIferylaDGala 10mg
SDP
Catalog No. 502967908
Encompass_Preferred
A fluorogenic substrate of α-galactosidase
Catalog No. 50-296-7908 Supplier Cayman Chemical Supplier No. 1655110 MG
A fluorogenic substrate of α-galactosidase