Learn More
Cayman Chemical 4-Methylumbelliferyl-α-D-Glucopyranoside, Purity: ≥95%, CAS: 17833-43-1, 100 mg
SDP
Catalog No. 502932698
Encompass_Preferred
A fluorogenic substrate for α-glucosidase; is cleaved by α-glucosidase to release the fluorescent moiety 4-MU; ex = 320 and 360 nm at low (1.97-6.72) and high pH (7.12-10.3), respectively; em = 445 to 455 nm, increasing as pH decreases; has been used to quantify α-glucosidase activity in infant blood spot samples in the diagnosis of Fabry and Pompe diseases
Catalog No. 50-293-2698 Supplier Cayman Chemical Supplier No. 26810100 MG
A fluorogenic substrate for α-glucosidase; is cleaved by α-glucosidase to release the fluorescent moiety 4-MU; ex = 320 and 360 nm at low (1.97-6.72) and high pH (7.12-10.3), respectively; em = 445 to 455 nm, increasing as pH decreases; has been used to quantify α-glucosidase activity in infant blood spot samples in the diagnosis of Fabry and Pompe diseases