Learn More
Cayman Chemical 4-Methylumbelliferyl-α-L-Fucopyranoside, Purity: ≥98%, CAS: 54322-38-2, 10 mg
SDP
Catalog No. 502916984
Encompass_Preferred
A fluorogenic substrate for α-L-fucosidase; 4-ΜU is released upon enzymatic cleavage by α-L-fucosidase and its fluorescence can be used to quantify α-L-fucosidase activity; ex/em = 350/440 nm, respectively
Catalog No. 50-291-6984 Supplier Cayman Chemical Supplier No. 3955210 MG
A fluorogenic substrate for α-L-fucosidase; 4-ΜU is released upon enzymatic cleavage by α-L-fucosidase and its fluorescence can be used to quantify α-L-fucosidase activity; ex/em = 350/440 nm, respectively