Learn More
Cayman Chemical 4-Methylumbelliferyl-N-acetyl-α-D-Neuraminic Acid (sodium salt), Purity: ≥98%, CAS: 76204-02-9, 10 mg
SDP
Catalog No. 502967183
Encompass_Preferred
A fluorogenic substrate of neuraminidases; cleaved by neuraminidases to release the fluorescent moiety 4-MU; has been used to quantify neuraminidase activity in mammalian cells and clinical isolates of influenza virus
Catalog No. 50-296-7183 Supplier Cayman Chemical Supplier No. 1662010 MG
A fluorogenic substrate of neuraminidases; cleaved by neuraminidases to release the fluorescent moiety 4-MU; has been used to quantify neuraminidase activity in mammalian cells and clinical isolates of influenza virus