Learn More
Cayman Chemical 4-Methylumbelliferyl β-D-N,N'-diacetylchitobioside, Purity: ≥98%, CAS: 53643-12-2, 10 mg
SDP
Catalog No. 502940970
Encompass_Preferred
A fluorogenic substrate for chitinases and chitobiosidases; cleaved by chitinases and chitobiosidases to release the fluorescent moiety 4-ΜU; ex/em = 320 and 360 at low (1.97-6.72) and high pH (7.12-10.3), respectively/445-455 nm, increasing as pH decreases
Catalog No. 50-294-0970 Supplier Cayman Chemical Supplier No. 2773010 MG
A fluorogenic substrate for chitinases and chitobiosidases; cleaved by chitinases and chitobiosidases to release the fluorescent moiety 4-ΜU; ex/em = 320 and 360 at low (1.97-6.72) and high pH (7.12-10.3), respectively/445-455 nm, increasing as pH decreases