Learn More
Chemglass Life Sciences Chemglass 4 mL (1 Dram) Sample Vial, 15 mm OD x 45 mm, 13-425 Thread, PTFE-Lined Black Cap, Lab-Pac (144)
SDP
Catalog No. NC3050257
Encompass_Preferred
Chemglass 4 mL (1 dram) clear sample vial made from Type 1 borosilicate glass, supplied with PTFE-lined solid PTFE black caps in lab-pack quantities. Designed for sample storage and autosampler use, it offers chemical resistance and dimensional consistency.
  • Type 1 borosilicate glass construction for chemical resistance.
  • 4 mL (1 dram) capacity suitable for small-volume samples.
  • 15 mm external diameter and 45 mm height for autosampler compatibility.
  • 13-425 thread size fits standard screw caps.
  • Includes PTFE-lined solid PTFE black caps for leak resistance and chemical inertness.
  • Packaged in a lab-pac of 144 vials for bulk use.
Catalog No. NC3050257 Supplier Chemglass Life Sciences Supplier No. CG4900B01
Chemglass 4 mL (1 dram) clear sample vial made from Type 1 borosilicate glass, supplied with PTFE-lined solid PTFE black caps in lab-pack quantities. Designed for sample storage and autosampler use, it offers chemical resistance and dimensional consistency.
  • Type 1 borosilicate glass construction for chemical resistance.
  • 4 mL (1 dram) capacity suitable for small-volume samples.
  • 15 mm external diameter and 45 mm height for autosampler compatibility.
  • 13-425 thread size fits standard screw caps.
  • Includes PTFE-lined solid PTFE black caps for leak resistance and chemical inertness.
  • Packaged in a lab-pac of 144 vials for bulk use.