Learn More
Alkali Scientific 4mL Glass 15x45mm Flat Base 13 425 Screw Thread Vial with Label, 100/Pk, Clear
SDP
Catalog No. 5000662418
Encompass_Preferred
The Alkali Scientific™ 4 mL Glass Vial with Label is designed for reliable sample storage and handling in laboratory and analytical applications. Made from high quality clear glass, this vial provides excellent chemical resistance while allowing clear visibility of sample contents during preparation and analysis. The flat base design ensures stability during handling and when used with autosamplers, while the 13 425 screw thread finish allows compatibility with a wide range of standard caps and closures. Each vial includes a pre applied label to support clear sample identification and efficient laboratory tracking. With dimensions of 15 × 45 mm, the vial fits easily into common laboratory workflows and storage racks. Supplied in convenient packs of 100, these vials are suitable for chromatography, routine analytical testing, and general laboratory sample preparation and storage tasks.
Catalog No. 50-006-62418 Supplier Alkali Scientific Supplier No. CV4400
The Alkali Scientific™ 4 mL Glass Vial with Label is designed for reliable sample storage and handling in laboratory and analytical applications. Made from high quality clear glass, this vial provides excellent chemical resistance while allowing clear visibility of sample contents during preparation and analysis. The flat base design ensures stability during handling and when used with autosamplers, while the 13 425 screw thread finish allows compatibility with a wide range of standard caps and closures. Each vial includes a pre applied label to support clear sample identification and efficient laboratory tracking. With dimensions of 15 × 45 mm, the vial fits easily into common laboratory workflows and storage racks. Supplied in convenient packs of 100, these vials are suitable for chromatography, routine analytical testing, and general laboratory sample preparation and storage tasks.