Learn More
Waters Corp Waters Corporation Vial, LCGC Certified, Clear Glass, Screw Neck, 4 mL
Catalog No. 50782520 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These certified vials are HPLC-tested for cleanliness and manufactured to tight dimensional tolerances. Made from chemically inert glass, they feature a PTFE/Silicone septum for excellent resealing, multiple injections, and sample storage across a wide temperature range.
  • HPLC-tested for cleanliness, detecting minute chemical concentrations.
  • Each batch individually examined for cleanliness post-packaging.
  • Headspace GC test confirms silicone septa healing effectiveness.
  • Chemically inert glass prevents compromise of analytical results.
  • PTFE/Silicone septum offers excellent resealing for multiple injections and sample storage.
  • Septum operates within a -40°C to 200°C temperature range.
  • Universal screw cap ensures secure sample and solvent storage.
  • No special tools needed for assembly.
  • Chemically inert materials prevent analyte interference.
Catalog No. 50-782-520 Supplier Waters Corp Supplier No. 186000838C
These certified vials are HPLC-tested for cleanliness and manufactured to tight dimensional tolerances. Made from chemically inert glass, they feature a PTFE/Silicone septum for excellent resealing, multiple injections, and sample storage across a wide temperature range.
  • HPLC-tested for cleanliness, detecting minute chemical concentrations.
  • Each batch individually examined for cleanliness post-packaging.
  • Headspace GC test confirms silicone septa healing effectiveness.
  • Chemically inert glass prevents compromise of analytical results.
  • PTFE/Silicone septum offers excellent resealing for multiple injections and sample storage.
  • Septum operates within a -40°C to 200°C temperature range.
  • Universal screw cap ensures secure sample and solvent storage.
  • No special tools needed for assembly.
  • Chemically inert materials prevent analyte interference.