Learn More
Waters Corp Waters Autosampler Vial, LCGC Certified, Clear Glass, 4 mL, Screw Neck
Catalog No. 50782522 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These certified vials are tested for cleanliness by HPLC and held to the tightest dimensional tolerances in the industry, ensuring reliable analytical results. They are manufactured to the highest ISO 9001 standards and chemically tested and approved.
  • Tested for cleanliness by HPLC.
  • Held to tightest dimensional tolerances.
  • Manufactured to ISO 9001 standards.
  • Certified to be within dimensional tolerances for autosamplers.
  • Each batch tested after packaging for cleanliness and quality.
  • Screw-top mechanically retains the septum in place during piercing and produces a good seal.
  • Preslit PTFE/Silicone septa ideal for numerous injections and sample storage due to excellent resealing capabilities.
  • Operates at temperatures ranging from -40°C to 200°C.
  • Vial, cap, & septum are included.
Catalog No. 50-782-522 Supplier Waters Corp Supplier No. 186000839C
These certified vials are tested for cleanliness by HPLC and held to the tightest dimensional tolerances in the industry, ensuring reliable analytical results. They are manufactured to the highest ISO 9001 standards and chemically tested and approved.
  • Tested for cleanliness by HPLC.
  • Held to tightest dimensional tolerances.
  • Manufactured to ISO 9001 standards.
  • Certified to be within dimensional tolerances for autosamplers.
  • Each batch tested after packaging for cleanliness and quality.
  • Screw-top mechanically retains the septum in place during piercing and produces a good seal.
  • Preslit PTFE/Silicone septa ideal for numerous injections and sample storage due to excellent resealing capabilities.
  • Operates at temperatures ranging from -40°C to 200°C.
  • Vial, cap, & septum are included.