Learn More
Alkali Scientific 4mL High Recovery Vial Clear Glass 15x51mm Flat Base 13 425 Screw Thread, 100/Pk
SDP
Catalog No. 5000661931
Encompass_Preferred
The Alkali Scientific™ 4 mL High Recovery Vial is designed to maximize sample recovery while delivering reliable performance for analytical and laboratory applications. Manufactured from high quality clear glass, the vial offers excellent chemical resistance and clear visibility of contents. The high recovery design reduces residual sample loss, making it suitable for low volume or high value samples used in analytical testing. A flat base provides stability during handling and autosampler operation, while the 13 425 screw thread finish ensures compatibility with standard caps and closures. Supplied in convenient 100 pack quantities, these vials support routine analysis, chromatography workflows, and high throughput laboratory environments.
Catalog No. 50-006-61931 Supplier Alkali Scientific Supplier No. HRV04
The Alkali Scientific™ 4 mL High Recovery Vial is designed to maximize sample recovery while delivering reliable performance for analytical and laboratory applications. Manufactured from high quality clear glass, the vial offers excellent chemical resistance and clear visibility of contents. The high recovery design reduces residual sample loss, making it suitable for low volume or high value samples used in analytical testing. A flat base provides stability during handling and autosampler operation, while the 13 425 screw thread finish ensures compatibility with standard caps and closures. Supplied in convenient 100 pack quantities, these vials support routine analysis, chromatography workflows, and high throughput laboratory environments.