Learn More
Chemglass Life Sciences Vial, 4mL (1 Dram), Complete with Green Open Top SURE-Link Polypropylene Cap, TFE Septa, 15x45mm, 13-425 Thread
SDP
Catalog No. NC9977207
Encompass_Preferred
Reaction Vial, 4mL (1 Dram), Complete with Green Open Top Sure-Link Polypropylene Cap, TFE Septa, 15x45mm, 13-425 Thread
Catalog No. NC9977207 Supplier Chemglass Life Sciences Supplier No. CG490405
Reaction Vial, 4mL (1 Dram), Complete with Green Open Top Sure-Link Polypropylene Cap, TFE Septa, 15x45mm, 13-425 Thread