Learn More
KPM ANALYTICS NORTH AMERICA CORPORATION KPM Analytics 4 mL Sample Cup, Optimized For Automated Systems, Reduced Cross-Contamination Risk, 1000 pieces
Catalog No. NC3072663
Encompass
The KPM Analytics 4 mL Sample Cup is designed for use with SmartChem series analyzers, ensuring reliable sample handling in automated testing. This single-use cup minimizes cross-contamination, supporting accurate analytical results. Its high optical clarity and polystyrene material make it ideal for photometric analysis, while the 1000-pack offers cost-effective workflows for laboratories seeking efficient sample management.
Catalog No. NC3072663 Supplier KPM ANALYTICS NORTH AMERICA CORPORATION Supplier No. 127001801
The KPM Analytics 4 mL Sample Cup is designed for use with SmartChem series analyzers, ensuring reliable sample handling in automated testing. This single-use cup minimizes cross-contamination, supporting accurate analytical results. Its high optical clarity and polystyrene material make it ideal for photometric analysis, while the 1000-pack offers cost-effective workflows for laboratories seeking efficient sample management.