Learn More
Chemglass Life Sciences Chemglass Transport Tubes, 4 mL, Sterile, Caps Separated, Nested (200 tubes/bag, 1,000 tubes/case)
SDP
Catalog No. 502733398
Encompass_Preferred
Chemglass 4 mL sterile transport tubes with caps separated (nested) are designed for secure storage, transport, and freezing of biological, clinical, environmental, and food testing samples. Manufactured in clean-room conditions and sterilized for immediate use, they offer reliable sealing for air shipment and cryogenic storage.
  • Material: USP VI polypropylene tube and HDPE cap.
  • Packaged: 200 tubes per bag, 200 caps per bag; 1,000 tubes and 1,000 caps per case.
  • Sterile by electron beam sterilization with SAL = 10^-6.
  • Produced in a 100,000-level clean workshop; DNAse-free, RNase-free, pyrogen-free, endotoxin-free.
  • Temperature range approximately -196°C to 121°C.
  • Sealing performance prevents leakage at negative pressure of 95 KPa, suitable for air transport.
Catalog No. 50-273-3398 Supplier Chemglass Life Sciences Supplier No. CGN474224
Chemglass 4 mL sterile transport tubes with caps separated (nested) are designed for secure storage, transport, and freezing of biological, clinical, environmental, and food testing samples. Manufactured in clean-room conditions and sterilized for immediate use, they offer reliable sealing for air shipment and cryogenic storage.
  • Material: USP VI polypropylene tube and HDPE cap.
  • Packaged: 200 tubes per bag, 200 caps per bag; 1,000 tubes and 1,000 caps per case.
  • Sterile by electron beam sterilization with SAL = 10^-6.
  • Produced in a 100,000-level clean workshop; DNAse-free, RNase-free, pyrogen-free, endotoxin-free.
  • Temperature range approximately -196°C to 121°C.
  • Sealing performance prevents leakage at negative pressure of 95 KPa, suitable for air transport.