Learn More
Nova Biostorage 4mL Internal Threaded , Tubes only, no Caps
SDP
Catalog No. NC2745853
Encompass
A massive working volume in a 48-well format characterizes the 4.00ml cryo tubes from Micronic. The sample storage vials are ideal for storing or transporting larger volumes of liquid or solid biological samples. Incorporating an internal thread screw cap with silicon O-ring – Micronic 4.00ml tubes provide optimal sealing enabling high integrity storage at ultra-low temperatures.
Catalog No. NC2745853 Supplier Nova Biostorage Supplier No. MP52440
A massive working volume in a 48-well format characterizes the 4.00ml cryo tubes from Micronic. The sample storage vials are ideal for storing or transporting larger volumes of liquid or solid biological samples. Incorporating an internal thread screw cap with silicon O-ring – Micronic 4.00ml tubes provide optimal sealing enabling high integrity storage at ultra-low temperatures.