Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Globe Scientific 4mL Tube with Screwcap and O-ring for Access™ Analyzer

Catalog No. 21200110 Shop All Globe Scientific Products
Change view
Click to view available options
Quantity:
1000/Cs.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-200-110 1000/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-200-110 Supplier Globe Scientific Supplier No. 6144
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Self-standing polypropylene tube/vial with cap for use with Beckman™ Access™ Analyzer

Serum transport tube designed for use with the Beckman Access™ analyzer.

  • Volume: 4.0 mL
  • Material: Polypropylene (PP)
  • Bottom Style: Round bottom
  • Design: Self-standing
  • Cap Style: Separate screw cap with O-ring

Specifications

Description Tube and cap are polypropylene
Color Clear
Material Polypropylene
Volume (Metric) 4 mL
Closure Type Screw Cap (Separate)
Quantity 1000/Cs.
Marking Spot No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.