Learn More
Ace Glass, Inc. Stopcock adapter, 28/15 outer top and bottom, 4mm glass plug
SDP
Catalog No. 50978194
Encompass_Preferred
Stopcock adapter, 28/15 outer top and bottom, 4mm glass plug
Catalog No. 50-978-194 Supplier Ace Glass, Inc. Supplier No. 384052
Stopcock adapter, 28/15 outer top and bottom, 4mm glass plug