Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Metrohm USA 4MM -2MM PLUG ADP RED
SDP

Catalog No. NC0283404 Shop All Metrohm USA Products
Encompass

For connecting plug B (4 mm) to 2 mm socket.

Catalog No. NC0283404 Supplier Metrohm USA Supplier No. 62103130
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For connecting plug B (4 mm) to 2 mm socket.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.