Learn More
Chemglass Life Sciences Glassblowers Condenser Coil, 4mm OD, Coil Dimension 14mm OD X 75mm Length
SDP
Catalog No. CG62701
Encompass_Preferred
Glassblowers Condenser Coil, 4mm OD, Coil Dimension 14mm OD X 75mm Length. For use in construction of spiral or Graham style condensers. Coils can be sealed together to form longer lengths.
Catalog No. CG62701 Supplier Chemglass Life Sciences Supplier No. CG62701
Glassblowers Condenser Coil, 4mm OD, Coil Dimension 14mm OD X 75mm Length. For use in construction of spiral or Graham style condensers. Coils can be sealed together to form longer lengths.