Learn More
Chemglass Life Sciences Stopcock, Double Oblique Bore, Glass Plug, 4mm Bore, 16.2/56 Plug Size, 10mm OD Sidearms
SDP
Catalog No. CG41402
Encompass_Preferred
Stopcock, Double Oblique Bore, Glass Plug, 4mm Bore, 16.2/56 Plug Size, 10mm OD Sidearms. Stopcock has a solid glass plug having parallel diagonal bores. Both sizes are supplied complete with metal retaining clip CG-423. The CG-414-02 is supplied with a gum washer.
Catalog No. CG41402 Supplier Chemglass Life Sciences Supplier No. CG41402
Stopcock, Double Oblique Bore, Glass Plug, 4mm Bore, 16.2/56 Plug Size, 10mm OD Sidearms. Stopcock has a solid glass plug having parallel diagonal bores. Both sizes are supplied complete with metal retaining clip CG-423. The CG-414-02 is supplied with a gum washer.