Learn More
Wilmad Labglass EPR Sample Tube, Thin Wall Quartz, 4mm
Catalog No. NC0945396
Encompass_Preferred
4 mm Thin Wall Quartz EPR Sample Tube 250 mm L
Catalog No. NC0945396 Supplier Wilmad Labglass Supplier No. 707SQ250M
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
4 mm Thin Wall Quartz EPR Sample Tube 250 mm L