Learn More
BIOSMITH Consumables. 4mm gap electroporation cuvette. Fits all commercial electroporators. Single packaged. 50 per pack
Catalog No. NC2980206
Encompass
Consumables. 4mm gap electroporation cuvette. Fits all commercial electroporators. Single packaged. 50 per pack
Catalog No. NC2980206 Supplier BIOSMITH Supplier No. 72004
Consumables. 4mm gap electroporation cuvette. Fits all commercial electroporators. Single packaged. 50 per pack