Learn More
Chemglass Life Sciences Valve, Chem-Vac, Glass Barrel Only, Without Sidearm, 0-4mm Bore, 13mm OD Barrel
SDP
Catalog No. CG99801
Encompass_Preferred
Valve, Chem-Vac, Glass Barrel Only, Without Sidearm, 0-4mm Bore, 13mm OD Barrel. Glass barrel only, without sidearm, for all CHEM-VAC valves. Heavy duty rounded coarse thread and precision formed tapered seat.PLEASE NOTE: THE 0-20mm BORE SIZE VALVE IS OUR PREVIOUS STYLE CG-500 SERIES VALVE WITH COARSE THREAD.
Catalog No. CG99801 Supplier Chemglass Life Sciences Supplier No. CG99801
Valve, Chem-Vac, Glass Barrel Only, Without Sidearm, 0-4mm Bore, 13mm OD Barrel. Glass barrel only, without sidearm, for all CHEM-VAC valves. Heavy duty rounded coarse thread and precision formed tapered seat.PLEASE NOTE: THE 0-20mm BORE SIZE VALVE IS OUR PREVIOUS STYLE CG-500 SERIES VALVE WITH COARSE THREAD.