Learn More
Chemglass Life Sciences Glass Shell Only: 4mm Bore, 15.2/30 Plug Size, 10 & 12mm OD Sidearms
SDP
Catalog No. 501532001
Encompass_Preferred
Glass Shell Only: 4mm Bore, 15.2/30 Plug Size, 10 & 12mm OD Sidearms. Similar to CG-424 but with one arm of the stopcock being of a larger diameter. Larger arm facilitates construction of stronger apparatus such as separatory funnels and burets. Available complete with 1:5 taper PTFE plug or glass shell only. FOR REPLACEMENT PLUGS, SEE CG-424 LISTING
Catalog No. 50-153-2001 Supplier Chemglass Life Sciences Supplier No. CGB42822
Glass Shell Only: 4mm Bore, 15.2/30 Plug Size, 10 & 12mm OD Sidearms. Similar to CG-424 but with one arm of the stopcock being of a larger diameter. Larger arm facilitates construction of stronger apparatus such as separatory funnels and burets. Available complete with 1:5 taper PTFE plug or glass shell only. FOR REPLACEMENT PLUGS, SEE CG-424 LISTING