Learn More
Chemglass Life Sciences Stopcock, Straight Bore, Glass Plug, 1-Arm Larger, 4mm Bore, 17/40 Plug Size, 10mm & 12mm OD Sidearms, #4 Clip Size
SDP
Catalog No. 501531645
Encompass_Preferred
Stopcock, Straight Bore, Glass Plug, 1-Arm Larger, 4mm Bore, 17/40 Plug Size, 10mm & 12mm OD Sidearms, #4 Clip Size. Similar to CG-400, but with one arm of the stopcock being of a larger diameter. Larger arm facilitates construction of stronger apparatus such as separatory funnels and burets. Supplied complete with the listed size metal retaining clip CG-423.
Catalog No. 50-153-1645 Supplier Chemglass Life Sciences Supplier No. CG40202
Stopcock, Straight Bore, Glass Plug, 1-Arm Larger, 4mm Bore, 17/40 Plug Size, 10mm & 12mm OD Sidearms, #4 Clip Size. Similar to CG-400, but with one arm of the stopcock being of a larger diameter. Larger arm facilitates construction of stronger apparatus such as separatory funnels and burets. Supplied complete with the listed size metal retaining clip CG-423.