Learn More
Ace Glass, Inc. 4mm hi vacuum stopcock HLW double oblique
SDP
Catalog No. 50979742
Encompass_Preferred
4mm precision-grade, hi vacuum stopcock, with double oblique bore hollow plug. Plug has a hole to permit evacuation of the lower chamber, and prevents leakage through the bottom.
Catalog No. 50-979-742 Supplier Ace Glass, Inc. Supplier No. 821206
4mm precision-grade, hi vacuum stopcock, with double oblique bore hollow plug. Plug has a hole to permit evacuation of the lower chamber, and prevents leakage through the bottom.