Learn More
Ace Glass, Inc. 4mm hi vacuum stopcock SLD double oblique
SDP
Catalog No. 50974479
Encompass_Preferred
4mm precision-grade, hi vacuum stopcock, with double oblique bore solid plug. Plug has a hole to permit evacuation of the lower chamber, and prevents leakage through the bottom.
Catalog No. 50-974-479 Supplier Ace Glass, Inc. Supplier No. 821228
4mm precision-grade, hi vacuum stopcock, with double oblique bore solid plug. Plug has a hole to permit evacuation of the lower chamber, and prevents leakage through the bottom.