Learn More
Ace Glass, Inc. 4MM HI VACUUM STOPCOCK HLW 4MM HI VACUUM STOPCOCK HLW
SDP
Catalog No. NC0827485
Encompass_Preferred
Oblique bore stopcock with vacuum cup at bottom. Available with a hollow or solid plug.
Catalog No. NC0827485 Supplier Ace Glass, Inc. Supplier No. 820906
Oblique bore stopcock with vacuum cup at bottom. Available with a hollow or solid plug.