Learn More
Ace Glass, Inc. 4MM ID GRADED SEALS 4MM ID GRADED SEALS
SDP
Catalog No. 50974590
Encompass
4mm quartz graded seals for connecting Pyrex or Kimax borosilicate glass to Quartz glass. Quartz length is 7.6cm, and borosilicate length is 10.2cm.
Catalog No. 50-974-590 Supplier Ace Glass, Inc. Supplier No. 845504
4mm quartz graded seals for connecting Pyrex or Kimax borosilicate glass to Quartz glass. Quartz length is 7.6cm, and borosilicate length is 10.2cm.