Learn More
Chemglass Life Sciences 4mm Diameter Quartz Rod, 48in Length
SDP
Catalog No. CGQ080103
Encompass_Preferred
4mm Diameter Quartz Rod, 48in Length. General Electric Type 214 clear fused quartz rod. Rod is stocked in 4ft lengths. PLEASE NOTE: PRICE LISTED IS PER LENGTH OF 4FT ROD.
Catalog No. CGQ080103 Supplier Chemglass Life Sciences Supplier No. CGQ080103
4mm Diameter Quartz Rod, 48in Length. General Electric Type 214 clear fused quartz rod. Rod is stocked in 4ft lengths. PLEASE NOTE: PRICE LISTED IS PER LENGTH OF 4FT ROD.