Learn More
Ace Glass, Inc. 4MM S/C 45/50M COLUMN 4MM S/C 45/50M COLUMN
SDP
Catalog No. 50975466
Encompass
Catalyst Column with two 4mm stopcocks and a 45/50 inner joint for the ACE-Burlitch system. The ACE-Burlitch inert atmosphere system is designed for maximum convenience and versatility in the handling of air-sensitive compounds in conjunction with ACE no-air labware, or suitiably adapted conventional glassware.
Catalog No. 50-975-466 Supplier Ace Glass, Inc. Supplier No. 781834
Catalyst Column with two 4mm stopcocks and a 45/50 inner joint for the ACE-Burlitch system. The ACE-Burlitch inert atmosphere system is designed for maximum convenience and versatility in the handling of air-sensitive compounds in conjunction with ACE no-air labware, or suitiably adapted conventional glassware.