Learn More
Ace Glass, Inc. 4mm stopcock straight bore PTFE plug
SDP
Catalog No. 50976121
Encompass_Preferred
4mm straight bore stopcock with polished barrels and 1:5 PTFE plug.
Catalog No. 50-976-121 Supplier Ace Glass, Inc. Supplier No. 813808
4mm straight bore stopcock with polished barrels and 1:5 PTFE plug.