Learn More
Ace Glass, Inc. 4MM STOPCOCK (GB)
SDP
Catalog No. 50979700
Encompass_Preferred
4mm Newman type stopcock is used with a distilling head to enable the operator to determine reflux ratios and take-offs. Process is described in Industrial and Engineering Chemistry, Volume 14, No. 11, Page 902, Nov. 15, 1942.
Catalog No. 50-979-700 Supplier Ace Glass, Inc. Supplier No. 815206
4mm Newman type stopcock is used with a distilling head to enable the operator to determine reflux ratios and take-offs. Process is described in Industrial and Engineering Chemistry, Volume 14, No. 11, Page 902, Nov. 15, 1942.