Learn More
Ace Glass, Inc. 4MM STOPCOCK GB 4MM STOPCOCK GB
SDP
Catalog No. 50976116
Encompass_Preferred
Glass 4mm straight bore stopcock with a glass plug and a tapered flange, which insures maximum strength at the seal. The bore of the side arms is equal to or slightly larger than the bore of the plug.
Catalog No. 50-976-116 Supplier Ace Glass, Inc. Supplier No. 813708
Glass 4mm straight bore stopcock with a glass plug and a tapered flange, which insures maximum strength at the seal. The bore of the side arms is equal to or slightly larger than the bore of the plug.