Learn More
Chemglass Life Sciences Stopcock, Straight Bore, PTFE Metering Plug, 4mm Bore, 15.2/30 Plug Size, 10mm OD Sidearms
SDP
Catalog No. 50873717
Encompass_Preferred
Stopcock, Straight Bore, PTFE Metering Plug, 4mm Bore, 15.2/30 Plug Size, 10mm OD Sidearms. A 1:5 taper PTFE plug having a threaded needle valve to allow the adjustment of flow through the bore of the plug.
Catalog No. 50-873-717 Supplier Chemglass Life Sciences Supplier No. CG43402
Stopcock, Straight Bore, PTFE Metering Plug, 4mm Bore, 15.2/30 Plug Size, 10mm OD Sidearms. A 1:5 taper PTFE plug having a threaded needle valve to allow the adjustment of flow through the bore of the plug.