Learn More
Chemglass Life Sciences Valve, Chem-Vac, Glass Portion Only, 180°, 0-4mm Bore, 13mm OD Barrel, 9mm X 1.5W Sidearm
SDP
Catalog No. CG90401
Encompass_Preferred
Valve, Chem-Vac, Glass Portion Only, 180¿, 0-4mm Bore, 13mm OD Barrel, 9mm X 1.5W Sidearm. CHEM-VAC valve glass portion only. Valve has two sidearms sealed to the barrel at a 180¿ angle to one another and on the same plane. Heavy duty rounded coarse thread and precision formed tapered seat. Dimensions as listed.
Catalog No. CG90401 Supplier Chemglass Life Sciences Supplier No. CG90401
Valve, Chem-Vac, Glass Portion Only, 180¿, 0-4mm Bore, 13mm OD Barrel, 9mm X 1.5W Sidearm. CHEM-VAC valve glass portion only. Valve has two sidearms sealed to the barrel at a 180¿ angle to one another and on the same plane. Heavy duty rounded coarse thread and precision formed tapered seat. Dimensions as listed.