Learn More
Chemglass Life Sciences Valve, Chem-Vac, Glass Portion Only, Low Hold Up, 0-4mm Bore, 13mm OD Barrel, 9mm X 1.5W Sidearm
SDP
Catalog No. CG90501
Encompass_Preferred
Valve, Chem-Vac, Glass Portion Only, Low Hold Up, 0-4mm Bore, 13mm OD Barrel, 9mm X 1.5W Sidearm. CHEM-VAC valve glass portion only. Valve has two sidearms sealed to the barrel at a 180¿ angle to one another and bent to minimize hold up of fluids. Heavy duty rounded coarse thread and precision formed tapered seat. Dimensions as listed.
Catalog No. CG90501 Supplier Chemglass Life Sciences Supplier No. CG90501
Valve, Chem-Vac, Glass Portion Only, Low Hold Up, 0-4mm Bore, 13mm OD Barrel, 9mm X 1.5W Sidearm. CHEM-VAC valve glass portion only. Valve has two sidearms sealed to the barrel at a 180¿ angle to one another and bent to minimize hold up of fluids. Heavy duty rounded coarse thread and precision formed tapered seat. Dimensions as listed.