Learn More
Ace Glass, Inc. PTFE 4mm tube compression fitting to 1/8in male NPT
SDP
Catalog No. 50977672
Encompass
4mm PTFE tube compression fitting to 1/8 male NPT. All wetted surfaces are virgin PTFE. Nuts and gripper are PVDF.
Catalog No. 50-977-672 Supplier Ace Glass, Inc. Supplier No. 1270912
4mm PTFE tube compression fitting to 1/8 male NPT. All wetted surfaces are virgin PTFE. Nuts and gripper are PVDF.