Learn More
Cayman Chemical 4-methyl-α-Pyrrolidinobutiophenone (hydrochloride), Purity: ≥98%, CAS: 1214-15-9, 50 mg
SDP
Catalog No. 502894241
Encompass_Preferred
A synthetic cathinone with psychoactive properties which has recently been identified in party pills and powders; intended for forensic and research applications
Catalog No. 50-289-4241 Supplier Cayman Chemical Supplier No. 900119050 MG
A synthetic cathinone with psychoactive properties which has recently been identified in party pills and powders; intended for forensic and research applications