Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hach Company BioTector B3500 Reagent, 4.0 N Sodium Hydroxide (NaOH), 20 L

Catalog No. NC3718293 Shop All Hach Company Products
Encompass

Hach BioTector B3500 reagent, 4.0 N sodium hydroxide, 20 L, is a concentrated alkaline solution formulated for use as a reagent in total organic carbon (TOC) analysis. It is supplied in a 20-liter container appropriate for instrument reagent requirements. Consult the product SDS for handling, storage, and shipping information.

  • Concentration: 4.0 N sodium hydroxide.
  • Volume: 20 L container.
  • Application: reagent for TOC analysis.
  • Intended for use with compatible TOC analyzers and reagent systems.
  • Storage: keep container tightly closed, protect from moisture, store in a cool, well-ventilated place.
  • Safety: subject to hazardous shipping; consult SDS for handling and transport.

Catalog No. NC3718293 Supplier Hach Company Supplier No. 2038062
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Hach BioTector B3500 reagent, 4.0 N sodium hydroxide, 20 L, is a concentrated alkaline solution formulated for use as a reagent in total organic carbon (TOC) analysis. It is supplied in a 20-liter container appropriate for instrument reagent requirements. Consult the product SDS for handling, storage, and shipping information.

  • Concentration: 4.0 N sodium hydroxide.
  • Volume: 20 L container.
  • Application: reagent for TOC analysis.
  • Intended for use with compatible TOC analyzers and reagent systems.
  • Storage: keep container tightly closed, protect from moisture, store in a cool, well-ventilated place.
  • Safety: subject to hazardous shipping; consult SDS for handling and transport.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.