Learn More
Cayman Chemical 4ndrosten11ol3 17dIon 10mg
SDP
Catalog No. 502992551
Encompass_Preferred
A non-androgenic steroid; metabolized to various androgenic 5α-reduced C19 steroids in LNCaP cells
Catalog No. 50-299-2551 Supplier Cayman Chemical Supplier No. 3041010 MG
A non-androgenic steroid; metabolized to various androgenic 5α-reduced C19 steroids in LNCaP cells