Learn More
Cayman Chemical 4-Androsten-11β-ol-3,17-dione, Purity: ≥98%, CAS: 382-44-5, 50 mg
SDP
Catalog No. 502990839
Encompass_Preferred
A non-androgenic steroid; metabolized to various androgenic 5α-reduced C19 steroids in LNCaP cells
Catalog No. 50-299-0839 Supplier Cayman Chemical Supplier No. 3041050 MG
A non-androgenic steroid; metabolized to various androgenic 5α-reduced C19 steroids in LNCaP cells