Learn More
Cayman Chemical 4-Nitrophenyl α-L-rhamnopyranoside, Purity: ≥98%, CAS: 18918-31-5, 50 mg
SDP
Catalog No. 502939975
Encompass_Preferred
A colorimetric naringinase substrate; upon hydrolysis by the α-rhamnosidase subunit of naringinase, 4-nitrophenol is released which can be quantified by colorimetric detection at 405 nm as a measure of naringinase α-rhamnosidase activity; has been used in the detection of naringinase α-rhamnosidase activity in various bacteria
Catalog No. 50-293-9975 Supplier Cayman Chemical Supplier No. 2919550 MG
A colorimetric naringinase substrate; upon hydrolysis by the α-rhamnosidase subunit of naringinase, 4-nitrophenol is released which can be quantified by colorimetric detection at 405 nm as a measure of naringinase α-rhamnosidase activity; has been used in the detection of naringinase α-rhamnosidase activity in various bacteria