Learn More
Cayman Chemical 4-Nitrophenyl β-D-Cellobioside, Purity: ≥98%, CAS: 3482-57-3, 50 mg
SDP
Catalog No. 502993725
Encompass_Preferred
An enzyme substrate; has been used to characterize function of endoglucanase 1 from T. harzianum, β-glucosidase from T. xylanolyticum, and GH3 aryl β-glucosidases from A. oryzae
Catalog No. 50-299-3725 Supplier Cayman Chemical Supplier No. 2337850 MG
An enzyme substrate; has been used to characterize function of endoglucanase 1 from T. harzianum, β-glucosidase from T. xylanolyticum, and GH3 aryl β-glucosidases from A. oryzae