Learn More
Cayman Chemical 4-Nitrophenyl β-D-Glucuronide, Purity: ≥98%, CAS: 10344-94-2, 100 mg
SDP
Catalog No. 502917198
Encompass_Preferred
A colorimetric β-glucuronidase substrate; upon enzymatic cleavage, p-nitrophenol is released, which can be quantified by colorimetric detection at 405 nm as a measure of β-glucuronidase activity
Catalog No. 50-291-7198 Supplier Cayman Chemical Supplier No. 38756100 MG
A colorimetric β-glucuronidase substrate; upon enzymatic cleavage, p-nitrophenol is released, which can be quantified by colorimetric detection at 405 nm as a measure of β-glucuronidase activity