Learn More
Cayman Chemical 4NItrophenyl DXylopyrnos 100mg
SDP
Catalog No. 502918703
Encompass_Preferred
A chromogenic β-xylosidase substrate; upon hydrolysis by β-xylosidase, 4-nitrophenol is released, which can be quantified by colorimetric detection at 405 nm as a measure of β-xylosidase activity
Catalog No. 50-291-8703 Supplier Cayman Chemical Supplier No. 38883100 MG
A chromogenic β-xylosidase substrate; upon hydrolysis by β-xylosidase, 4-nitrophenol is released, which can be quantified by colorimetric detection at 405 nm as a measure of β-xylosidase activity