Learn More
Cayman Chemical 4-Nitrophenyl-N-acetyl-α-D-galactosaminide, Purity: ≥98%, CAS: 23646-68-6, 10 mg
SDP
Catalog No. 502965335
Encompass_Preferred
A chromogenic substrate for N-acetyl-D-galactosaminidase that generates a yellow solution upon cleavage
Catalog No. 50-296-5335 Supplier Cayman Chemical Supplier No. 1675510 MG
A chromogenic substrate for N-acetyl-D-galactosaminidase that generates a yellow solution upon cleavage