Learn More
Cayman Chemical 4NItrophenylNacetylDgl 100mg
SDP
Catalog No. 502941445
Encompass_Preferred
A chromogenic substrate for N-acetyl-β-glucosaminidase; hydrolysis of 4-nitrophenyl-N-acetyl-β-D-glucosaminide by N-acetyl-β-glucosaminidase releases 4-nitrophenol, which can be quantified by colorimetric detection at 405 nm as a measure of N-acetyl-β-glucosaminidase activity; has been used to quantify the activity of N-acetyl-β-D-glucosaminidase in human serum and urine in parenterally fed patients
Catalog No. 50-294-1445 Supplier Cayman Chemical Supplier No. 28954100 MG
A chromogenic substrate for N-acetyl-β-glucosaminidase; hydrolysis of 4-nitrophenyl-N-acetyl-β-D-glucosaminide by N-acetyl-β-glucosaminidase releases 4-nitrophenol, which can be quantified by colorimetric detection at 405 nm as a measure of N-acetyl-β-glucosaminidase activity; has been used to quantify the activity of N-acetyl-β-D-glucosaminidase in human serum and urine in parenterally fed patients