Learn More
Cayman Chemical 4-oxo-2-Nonenal, Purity: ≥98%, CAS: 103560-62-9, 5 mg
SDP
Catalog No. 502888408
Encompass_Preferred
A lipid peroxidation product; actively modifies histidine and lysine residues on proteins and causes protein cross-linking
Catalog No. 50-288-8408 Supplier Cayman Chemical Supplier No. 101855 MG
A lipid peroxidation product; actively modifies histidine and lysine residues on proteins and causes protein cross-linking